Tlačiť
Microcentrifuges, 5425/5425 R (General Lab Product)
Dodávateľ: EPPENDORF
5405000.310 5405000.719 5405000.174 5406000.119 5406000.313 5406000.518 5406000.712 5405000.117
521-2471EA 2680.55 EUR
521-2471 521-2479 521-2783 521-3351 521-3355 521-3359 521-3363 EPPE5405000.117
Microcentrifuges, 5425/5425 R (General Lab Product)
Centrifugy Mikrocentrifugy
This air-cooled centrifuge covers a range of very frequent applications. Focusing on standard applications, it is an essential piece of equipment for modern labs ensuring optimal separation. Ideal for high-throughput users expecting a top level of ergonomic comfort as well as high flexibility with 6 rotor options.
  • QuickLock® technology for ergonomic rotor lid locking
  • OptiBowl® design for quiet operation, even without a rotor lid
  • Soft touch one-finger lid closure for ergonomic operation
  • Increased software features (10 acceleration and braking ramps, 3
More Product Information