Tlačiť
Nalgene® Membrane Filters
Dodávateľ: Thermo Fisher Scientific
DS0205-4045 DS0210-4020 DS0210-4045 DS0215-4020 DS0215-4045 DS0205-6045
513-2052EA 171.04 EUR
513-2052 513-2101 513-2102 513-2151 513-2152 NALGDS0205-6045
Nalgene® Membrane Filters
Filtry Filtry membránové
Type 205: Cellulose nitrate, sterile. Certified to meet requirements of standard methods for water quality control for microbiological analysis, sterility tests, food and beverage testing. Individually wrapped.
Type 210: Cellulose acetate, non sterile, autoclavable. Filter for sterilisation of proteinaceous solutions. Low adsorption of proteins from serum or culture media. Resistant to alcohol and oil, can be used in short-term contact with methyl ethyl ketone, suitable for liquid scintillation counting. Must be wet when autoclaved.
Type 215: Nylon, hydrophilic, non sterile,More Product Information
Order Now
ŠPECIFIKÁCIA
Typ 205 210 210 215 215 205
Velikost póru 0,20 µm 0,45 µm 0,20 µm 0,45 µm 0,45 µm
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Objemový průtok vody >51 ml/min/cm² (1 bar) >21 ml/min/cm² (1 bar) >51 ml/min/cm² (1 bar) >19 ml/min/cm² (1 bar) >37 ml/min/cm² (1 bar) >51 ml/min/cm² (1 bar)
Maximální teplota 130 °C 120 °C 120 °C 125 °C 125 °C 130 °C
Certifikát ochrany životního prostředí NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement