Tlačiť
VWR®, Test Sieves, 305×60 mm, with Square Holes
Dodávateľ: VWR Collection
510-2929EA 310.84 EUR
510-2929 510-2930 510-2931 510-2932 510-2933 510-2934 510-2935 510-2936 510-2937 510-2938 510-3300 510-3301 510-3302 510-3303 510-3304 510-3305 510-3306 510-3307 510-3308 510-3309 510-3310 510-3311 510-3312 510-3313 510-3314 510-3315 510-3316 510-3317 510-3318 510-3319 510-3320 510-3321 510-3322 510-3323 510-3324 510-3325 510-3326 510-3327 510-3328 510-3329 510-3330
VWR®, Test Sieves, 305×60 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace oMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable