Tlačiť
VWR®, Test Sieves, 200×50 mm, with Square Holes
Dodávateľ: VWR Collection
510-0799EA 204.69 EUR
510-0799 510-0800 510-0801 510-0802 510-0803 510-0804 510-0805 510-0806 510-0807 510-0808 510-0809 510-0810 510-0811 510-0812 510-0813 510-0814 510-0815 510-0816 510-0817 510-0818 510-0819 510-0820 510-0821 510-0822 510-0823 510-0824 510-0825 510-0826 510-0827 510-0828 510-0829 510-0830 510-0831 510-0832 510-0833 510-0834 510-0835 510-0836 510-0837 510-0838 510-0839
VWR®, Test Sieves, 200×50 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace oMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable