Tlačiť
Thermoconductive tube racks for PCR tubes, microcentrifuge tubes, cryotubes, blood tubes and tall centrifuge tubes, Corning® CoolRack®
Dodávateľ: Corning
432036 432037 432038 432039 432040 432041 432042 432043 432044 432045 432046 432047 432048 432049 432050 432051 432052 432053 432054 432055 432056 432057 432058 432059 432060 432061 432062 432063 432064 432065 432066 432067 432068 432069 432070 432071 432072 432034 432035 432139
479-1932EA 81.31 EUR
479-1932 479-1853 479-1933 479-1934 479-1935 479-1936 479-1937 479-1938 479-1854 479-1855 479-1939 479-1856 479-1857 479-1858 479-1940 479-1859 479-1941 479-1942 479-1943 479-1944 479-1945 479-1946 479-1947 479-1948 479-1949 479-1950 479-1951 479-1952 CORN432064 479-1953 479-1954 479-1955 479-1956 479-1957 479-1958 479-1959 479-1960 479-1930 479-1931 CORN432139
Thermoconductive tube racks for PCR tubes, microcentrifuge tubes, cryotubes, blood tubes and tall centrifuge tubes, Corning® CoolRack®
Stojany Stojánky chladící
These thermoconductive modules provide reproducible and standardised temperature control to biological samples while on the bench.
  • Place CoolRack® or CoolSink™ modules on ice, dry ice, liquid nitrogen, water baths, heat blocks (–196 to +100 °C), or other vessels and they rapidly adapt to source temperature
Cool, (snap) freeze, or thaw samples consistently and reproducibly without directly inserting them into a temperature source to reduce variability and contamination. CoolRack® tube holders and CoolSink™ plate holders accommodate a wide variety ofMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable