Tlačiť
Storage canes for cryotubes CryoLine™, Nunc™
Dodávateľ: Thermo Fisher Scientific
378441
479-0021EA 55.88 EUR
479-0021
Storage canes for cryotubes CryoLine™, Nunc™
Slámky kryogenní
Aluminium storage canes for six cryotubes of 1,2 or 1,8 ml. Four cryotubes of 3,6 ml or three cryotubes of 4,5 ml.
  • Can be marked with colour code tabs
Length: 280 mm
Limited Time Sale
Select items below are currently on sale