Tlačiť

Indium(III) iodide, ultra dry ≥99.999% (metals basis)

Dodávateľ: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

47108.04 47108.09 47108.18
47108.04EA 94.53 EUR
47108.04 47108.09 47108.18
Indium(III) iodide, ultra dry ≥99.999% (metals basis)
Indium(III) iodide

Purity: 99.999% CAS No.: 13510-35-5 Molecular Formula: InI3 Formula Weight: 495.53 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.69 Sensitivity: Light Sensitive; Hygroscopic Appearance: Yellow to brown Melting Point: 210° UN No.: UN3260 MDL No.: MFCD00016151

Vzorec: I₃In
Mw: 495.53
Teplota varu: 500
Teplota tavenia: 210 °C
Hustota: 4.69
Skladovacia teplota: Ambient
Číslo MDL: MFCD00016151
Číslo CAS: 13510-35-5
EINECS: 236-839-6
OSN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR