Tlačiť
Laboratory Refrigerator without explosion-proof interior LABO-720
Dodávateľ: KIRSCH PHILIPP
10473
471-2546EA 7621.07 EUR
471-2546
Laboratory Refrigerator without explosion-proof interior LABO-720
Chladničky
Temperature range: 0 to 20 °C.
  • Recirculated air cooling
  • Key protected control panel
  • Automatic defrosting
  • Melt water evaporation
  • Minimum/maximum temperature memory
  • Digital display and micro-processor controlled
  • Optical and acoustical alarm
  • Free contact RS485 Interface