Tlačiť
Pharmaceutical refrigerator, MED-125
Dodávateľ: KIRSCH PHILIPP
10668
471-2535EA 2884.04 EUR
471-2535
Pharmaceutical refrigerator, MED-125
Chladničky
This undercounter refrigerator is ideal for the installation in wall units. Suitable for the storage of vaccines.
  • 3 drawers and 1 shelf
  • Recirculation air-cooling
  • Automatic defrosting; melt water evaporation
  • Digital temperature display
  • Optical and acoustic warning device including power failure alarm
  • Safety control prevents freezing
  • Potential free contact for alarm transmission
  • Lockable door
Power consumption: 125 Watt
Temperature range: 2 to 20 °C
Certifikácie: According to DIN 58345