Tlačiť

Viscosity standard, Specpure®, nominally 50cSt @ 40° and 7.3cSt @ 100°

Dodávateľ: Thermo Fisher Scientific

47013.AP
47013.APEA 143 EUR
47013.AP
Viscosity standard, Specpure®, nominally 50cSt @ 40° and 7.3cSt @ 100°
Standardy Standardy pro Měření Viskozity

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00084411

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR