Tlačiť
Desiccator cabinets, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F420640001 F420660000 F420660001
467-0062EA 1121.97 EUR
467-0062 467-0066 467-0067
Desiccator cabinets, SP Bel-Art
Exsikátory Boxy exsikační
These durable cabinets are made from 6,35 mm thick acrylic. The gasketed, air-tight doors are made from 9,5 mm thick acrylic with full length hinges and stainless steel twist locking latches. Available in clear or dark bronze acrylic.
  • Dry, dust-free storage for use in freezers, refrigerators, or on the benchtop
  • Supplied with perforated trays which ensure proper circulation within the cabinet
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable