Tlačiť
Shaking water baths, Hydro Series
Dodávateľ: LAUDA
L002908 L002909 L002910
462-0616EA 3620.67 EUR
462-0616 462-0617 462-0618
Shaking water baths, Hydro Series
Lázně Lázně vodní třepací
Shaking water baths for exactly reproducible temperatures. The working temperature range is 25 to 100 °C.
  • Easy-to-read LED display
  • Setting and displaying temperature values in increments of 0,1 °C
  • Membrane keypad with concise symbols
  • Digital setpoint adjustment
  • Continuously adjustable speed, load-independent, with gently controlled start-up
  • 1-point temperature controller
  • Over-temperature protection for operation with non-flammable liquids and low water level protection
  • Electronic monitoring of the temperature controller
  • Shaking device with special