Tlačiť
Digital block heaters, QBD and QBH
Dodávateľ: GRANT INSTRUMENTS
QBD1 QBD4 QBH2 QBD2
460-0270EA 1141.6 EUR
460-0270 460-0079 460-0272 460-0271
Digital block heaters, QBD and QBH
Termostaty blokové Termostaty blokové/ohřívací
Compact digital block heaters with choice of models for 1 or 2 or 4 interchangeable blocks. Units feature an LED temperature display, easy to use interactive user interface for fast and accurate set up. Variable high and low temperature alarms can be set to within 0,5 °C of the set point. Heaters are made from cool wall aluminium and chemically resistant plastic, have a low profile, a sloping fascia for ease of use, off-set adjustment for high accuracy operating temperatures and 0,1 °C display resolution.
  • QBD models have operating range of ambient +5 to 130 °C, QBH has an upper