Tlačiť
VWR® ARISTAR®, Interference Check B Nitric Acid 2% for ICP-MS
Dodávateľ: VWR Chemicals
456562KEA 422.59 EUR
456562K
VWR® ARISTAR®, Interference Check B Nitric Acid 2% for ICP-MS
Standardy Standardy kationické Standardy víceprvkové
11 Elements in 2% nitric acid solution.
11 Elements : 10 mg/L As, Cd, Se Zn; 20 mg/L Ag, C, Cr, Cu, Mn, Ni, V
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Certifikácie: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Informace o dodávce: Supplied with certificate of analysis.