Tlačiť
Stroncium 1.000 mg/l v zřed. kyselině dusičné (from Sr(NO3)2) ARISTAR® standard pro ICP
Dodávateľ: VWR Chemicals
456062SEA 165.83 EUR
456062S 456064U
Stroncium 1.000 mg/l v zřed. kyselině dusičné (from Sr(NO3)2) ARISTAR® standard pro ICP
Standardy Standardy kationické Standardy - stroncium
Sr(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable