Tlačiť
Niób 1.000 mg/l ve vodě se stopami kyseliny fluorovodíkové (from Nb2O5) ARISTAR® standard pro ICP
Dodávateľ: VWR Chemicals
455742YEA 368.92 EUR
455742Y 455744K
Niób 1.000 mg/l ve vodě se stopami kyseliny fluorovodíkové (from Nb2O5) ARISTAR® standard pro ICP
Standardy Standardy kationické Standardy - niob
Nb₂O₅ in H₂O tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.