Tlačiť
Mangan 10.000 mg/l v zřed. kyselině dusičné (from Mn(OAc)2) ARISTAR® standard pro ICP
Dodávateľ: VWR Chemicals
455652HEA 324.05 EUR
455652H 455654J
Mangan 10.000 mg/l v zřed. kyselině dusičné (from Mn(OAc)2) ARISTAR® standard pro ICP
Standardy Standardy kationické Standardy - mangan
Mn(C₂H₃O₂)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable