Tlačiť
Indium 10.000 mg/l v zřed. kyselině dusičné (from In) ARISTAR® standard pro ICP
Dodávateľ: VWR Chemicals
455492JEA 476.67 EUR
455492J
Indium 10.000 mg/l v zřed. kyselině dusičné (from In) ARISTAR® standard pro ICP
Standardy Standardy kationické Standardy - indium
In in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.