Tlačiť
Orbital shakers, KS 15 A and KS 15 A control
Dodávateľ: BUEHLER
6171000 6170000
444-9213EA 2370.12 EUR
444-9213 444-9212
Orbital shakers, KS 15 A and KS 15 A control
Třepačky Třepačky orbitální
Orbital shakers, which are suitable for continuous operation. The universal tray accepts max. 5×1000 ml or 9×500 ml flask clamps, Combifix rack system accommodates 4×1000 ml or 6×500 ml Erlenmeyer flasks. Variety of different vessels can be accommodated by choosing from the range of optional attachments.
  • KS 15 A has analogue control, KS 15 A control has digital display with a new programmable control with extended functionality
  • Continuous operation or timer operation from 0 to 120 min (KS 15 A), and continuous operation or programmable (KS 15 A control)
More Product Information