Tlačiť
Laboratory blenders, Seward Stomacher® 3500
Dodávateľ: FERMIONX SEWARD
3500/000 3500/001 3500/002
432-3130EA 7854.2 EUR
432-3130 432-0058 432-0059
Laboratory blenders, Seward Stomacher® 3500
Homogenizátory Mísiče laboratorní (blenders)
Stomacher® lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher® machines work by the action of twin reciprocating paddles on the sample contained inside a sterile plastic bag.
  • 3 year warranty, 10 year service and spare parts
  • Versatile and efficient blending and homogenisation of laboratory samples
  • Safe containment of sample in sterile plastic bag prevents cross-contamination
  • Machines do not need to be cleaned and sterilised between samples
  • No temperature increase
  • Sample volumes from 400 to 3500
More Product Information