Tlačiť

Polysorbate 80 NF, Multi-Compendial, J.T.Baker®

Dodávateľ: Avantor

Synonymá: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

Management of Change (MOC) category = R

4117-03EA 4186.26 EUR
4117-03 4117-04 4117-06 4117-12 4117-07 4117-02
Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Polysorbate 80

  • GMP manufactured product
  • Vegetable based


Suitable for use in the manufacture of parenteral dosage forms.

Teplota tavenia: –21 °C
Hustota: 1.062 g/cm³
Teplota vzplanutia: 113 °C (ot. pohárek)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00082107
Číslo CAS: 9005-65-6
EINECS: 500-019-9

Order Now

Výsledky špecifikačného testu

Meets E.P. Chemical Specifications ≤2.0
Meets B.P. Chemical Specifications 65 - 80
Meets N.F. Requirements Passes Test
Meets J.P. Chemical Specifications Passes Test
GMP Manufactured Product ≤0.25%
CAUTION: For Manufacturing, processing or repackaging 45 - 55
Bulk Pharmaceutical Chemical 1.06 - 1.09
NF - Acid Value ≤ 2.0
NF - Hydroxyl Value 65 - 80
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Residue on Ignition ≤ 0.25 %
NF - Saponification Value 45 - 55
NF - Specific Gravity at 25°C 1.06 - 1.09
NF - Viscosity at 25°C, cSt 300 - 500
NF - Water (H₂O) ≤ 3.0 %
NF - Peroxide Value ≤ 10.0
NF - Ethylene Oxide ≤ 1 ppm
NF - Dioxane ≤ 10 ppm
NF - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
NF - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
NF - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
NF - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
NF - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
NF - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
NF - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
EP - Acid Value ≤ 2.0
EP - Total Ash ≤ 0.25 %
EP - Hydroxyl Value 65 - 80
EP - Identification A Passes Test
EP - Identification D Passes Test
EP - Peroxide Value ≤ 10.0
EP - Ethylene Oxide ≤ 1 ppm
EP - Dioxan ≤ 10 ppm
EP - Saponification Value 45 - 55
EP - Water (H₂O) ≤ 3.0 %
EP/BP - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
EP/BP - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
EP/BP - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
EP/BP - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
EP/BP - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
EP/BP - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
EP/BP - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
Appearance Passes Test
JP - Acid Value ≤ 2.0
JP - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
JP - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
JP - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
JP - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
JP - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
JP - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
JP - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
JP - Dioxane ≤ 10 ppm
JP - Ethylene Oxide ≤ 1 ppm
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Hydroxyl Value, meq KOH/g 65 - 80
JP - Identification Passes Test
JP - Peroxide Value ≤ 10.0
JP - Residue on Ignition ≤ 0.25 %
JP - Water (H₂O) ≤ 3.0 %
JP - Saponification Value 45 - 55
Additional Tests - Color (Gardner) ≤ 7
Additional Tests - Odor (Faint) Passes Test
Additional Tests - Peroxide Value, meqO₂/kg ≤ 2.0
Additional Tests - Endotoxin Concentration (EU/mL) ≤ 10
Free Ethylene Oxide ≤ 1 ppm
1,4-Dioxane ≤ 5 ppm
Microbiological - Total Aerobic Microbial Count (cfu/g) ≤ 100
Microbiological - Escherichia Coli Passes Test
Microbiological - Pseudomonas aeruginosa Passes Test
Microbiological - Salmonella Passes Test
Microbiological - Staphylococcus aureus Passes Test
Microbiological - Total Yeast and Mold Count (cfu/g) ≤ 50
Residual Solvents - Ethylene Glycol, For Information Only
Residual Solvents - Acetic Acid, For Information Only
Residual Solvents - 2-Propanol, For Information Only
Ethylene glycol ≤ 0.10 %
Diethylene glycol ≤ 0.10 %
Vegetable Based
This product utilizes ingredients of non-animal origin and non-peanut origin.
Suitable for use in the manufacture of parenteral dosage forms.
Only Class 2 (1,4 Dioxane, Ethylene Glycol) and Class 3 (acetic acid, 2-propanol) solvents are likely to be present. Class 2 solvents are below the Option 1 limits and any Class 3 solvent is <0.5%.
Typical Oleic Acid Content, 77%
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.
Due to the anhydrous nature of this product, sodium oleate, a carboxylate salt/soap formed naturally in the process and which can be white to brown in color, can precipitate with time and may affect product viscosity.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR