Tlačiť

Síran amonný ≥99.5%, Ultrapure Bioreagent for molecular biology, for liquid chromatography, J.T.Baker®

Dodávateľ: Avantor
Varování

Synonymá: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

4027-02EA 138.59 EUR
4027-02 4027-06 4027-07 4027-09
Síran amonný ≥99.5%, Ultrapure Bioreagent for molecular biology, for liquid chromatography, J.T.Baker®
Síran amonný

For protein precipitation, liquid chromatography and molecular biology applications.

Vzorec: (NH₄)₂SO₄
Mw: 132.14
Teplota tavenia: 336…339 °C
Hustota: 1.78 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003391
Číslo CAS: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Výsledky špecifikačného testu

For Laboratory,Research,or Manufacturing Use ≥99.5%
Assay ((NH₄)₂SO₄) ≥99.5 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.0 - 6.0
Residue after Ignition ≤0.005 %
Absorbance of a 1M Solution - 260 nm ≤0.04
Absorbance of a 1M Solution - 280 nm ≤0.04
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Chloride (Cl) ≤3 ppm
Nitrate (NO₃) ≤10 ppm
Phosphate (PO₄) ≤3 ppm
Arsenic (As) ≤0.2 ppm
Trace Impurities - Cadmium (Cd) ≤2 ppm
Trace Impurities - Calcium (Ca) ≤2 ppm
Trace Impurities - Copper (Cu) ≤0.2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤0.1 ppm
Trace Impurities - Lithium (Li) ≤2 ppm
Trace Impurities - Magnesium (Mg) ≤2 ppm
Trace Impurities - Manganese (Mn) ≤0.6 ppm
Trace Impurities - Potassium (K) ≤2 ppm
Trace Impurities - Zinc (Zn) ≤0.5 ppm
Solution Test Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR