Tlačiť
Cell culture flasks with standard or filter caps, CELLSTAR®
Dodávateľ: Greiner Bio-One
658170_120 690160_200 660175_50 690170_200 658175_120 661175_40 661160_40 660160_50 690175_200
391-3106EA 242.74 EUR
391-3106 391-3103 391-3148 391-3104 391-3146 392-0013 392-0012 391-3109 391-3143
Cell culture flasks with standard or filter caps, CELLSTAR®
Baňky Baňky pro buněčné kultury
PS, sterile.
  • Improved cell adhesion due to physical surface treatment
  • Easy-open filter cap guarantees optimal gas exchange whilst minimising the risk of contamination
  • Stacking ridges on the top of the flask makes them easy to stack and ensures that they stand firmly in the incubator
  • Graduated on both sides for easier filling
  • Choice of flat or high profile design of the 175 cm² flask, allows for variability of media volume
These standard cell culture flasks are supplied with either standard screw caps or capillary pore membrane filter caps (poreMore Product Information