Tlačiť
Penicillin flask, DURAN®
Dodávateľ: DWK LIFE SCIENCES
215517106
391-0370EA 265.17 EUR
391-0370
Penicillin flask, DURAN®
Baňky Baňky kultivační
This penicillin flask is made from clear DURAN® borosilicate glass 3.3.
  • Neck is bent upwards which prevents the liquid culture medium from leaking
  • Due to the flat, large floor surface, the layer cover of microorganisms can grow evenly high
  • The large liquid surface ensures better aeration and thus faster growth of the microorganisms
Typically suitable for preparation of cultures in nutrient media.