Tlačiť
Constant Climate Chambers, KBF PRO Series
Dodávateľ: BINDER GMBH
9020-0439 9020-0440 9020-0441 9020-0442 9020-0443 9020-0444
390-1759EA 0 EUR
390-1759 390-1760 390-1761 390-1762 390-1763 390-1764
Constant Climate Chambers, KBF PRO Series
Komory testovací Komory klimatické
A powerful all-rounder among constant climate chambers: Equipped with all kinds of sophisticated technology, the KBF PRO with proven APT.line™ preheating chamber technology takes any material test in its stride – whether it takes place at -20 °C or under THB conditions at +85 °C and 85% relative humidity. Units have a stainless-steel inner chamber and temperature range of -20 to +100 °C and humidity range of 10% to 98% RH. The models offer flexible water supply options for all installation sites and consumes up to 40% less energy than its predecessor, the KMF model. Optional lightMore Product Information