Tlačiť
VWR® INCU-Line® Prime, Incubators with Natural Convection
Dodávateľ: VWR Collection
390-1346EA 2088.92 EUR
390-1346 390-1349 390-1350
VWR® INCU-Line® Prime, Incubators with Natural Convection
Inkubátory Inkubátory standardní
INCU-Line® microbiological incubators are designed to cultivate organisms at homogeneous temperatures. They have a temperature range of 5 °C above ambient to 100 °C and can, for instance, be used for microbiological tests, germ count determination and ageing/heated storage.
  • Multi functional microprocessor controller with 4,3" full colour touch screen
  • Five user program memory
  • Five-segment temperature-time profile, quick change of set temperature and set time during a running program
  • Scheduled program start (set of date and time) and loop function up to 255
More Product Information