Tlačiť

Chlorid zinečnatý, ultra dry ≥99.999% (metals basis)

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

Synonymá: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

Ampuled under argon

35726.06 35726.14 35726.22
35726.06EA 120.97 EUR
35726.06 35726.14 35726.22
Chlorid zinečnatý, ultra dry ≥99.999% (metals basis)
Chlorid zinečnatý

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: Powder, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Vzorec: ZnCl₂
Mw: 136.3
Teplota varu: 732 °C
Teplota tavenia: 290 °C
Hustota: 2.91
Skladovacia teplota: Ambient
Číslo MDL: MFCD00011295
Číslo CAS: 7646-85-7
EINECS: 231-592-0
OSN: 2331
ADR: 8 III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR