Tlačiť

Síran amonný ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™

Dodávateľ: Avantor Performance Materials
Varování

Synonymá: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

3512-65EA 3675.95 EUR
3512-65 3512-06 3512-12 3512-25 3512-28 3512-88
Síran amonný ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Síran amonný
Vzorec: (NH₄)₂SO₄
Mw: 132.14
Teplota tavenia: 336…339 °C
Hustota: 1.77 g/cm³
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003391
Číslo CAS: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Výsledky špecifikačného testu

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥99.0 %
ACS - Residue on Ignition ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - Phosphate (PO₄) ≤3 ppm
ACS - Iron (Fe) ≤5 ppm
Heavy Metals (as Pb) ≤3 ppm
Arsenic (As) ≤0.200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR