Tlačiť

n-Hexan ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™

Dodávateľ: Honeywell Chemicals
Nebezpečí

Synonymá: Hexyl hydride, Hexanen, Hexane

34859-45L 34859-7L 34859-1L 34859-2.5L 34859-4X2.5L 34859-6X1L 34859-200L
34859-45LEA 7730.42 EUR
34859-45L 34859-7L 34859-1L 34859-2.5L 34859-4X2.5L 34859-6X1L HONC34859-200L
n-Hexan ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
n-Hexan

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Vzorec: H₃C(CH₂)₄CH₃
Mw: 86.18
Teplota varu: 66…69 °C(1.013 hPa)
Teplota tavenia: –95 °C
Hustota: 0.660…0.668 g/cm³ (20 °C)
Teplota vzplanutia: 7.4 %(V)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00009520
Číslo CAS: 110-54-3
EINECS: 203-777-6
OSN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Assay (GC) Min. 97.00 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.010 %
Free acid (as CH₃COOH) Max. 0.001 %
Absorbance at 250 nm Max. 0.01
Absorbance at 225 nm Max. 0.1
Absorbance at 220 nm Max. 0.7
Absorbance at 210 nm Max. 0.3
Absorbance (JP)(Val.Ref) complying
IR spectrum complying
Peroxides JP complying
Vapor Pressure ~132 mmHg ( 20 °C)
Refractive Index n20/D 1.375 (lit.)
Vapor Density ~3 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR