Tlačiť

n-Hexan 95, CHROMASOLV™ Pest_Residual, Riedel-de Haën™

Dodávateľ: Honeywell Chemicals
Nebezpečí

Synonymá: Hexyl hydride, Hexanen, Hexane

34484-7L 34484-18L 34484-45L 34484-1L 34484-2.5L 34484-4X2.5L
34484-7LEA 711.8 EUR
34484-7L 34484-18L 34484-45L 34484-1L 34484-2.5L 34484-4X2.5L
n-Hexan 95, CHROMASOLV™ Pest_Residual, Riedel-de Haën™
n-Hexan

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Vzorec: H₃C(CH₂)₄CH₃
Mw: 86.18
Teplota varu: 66…69 °C(1.013 hPa)
Teplota tavenia: –95 °C
Hustota: 0.660…0.668 g/cm³ (20 °C)
Teplota vzplanutia: 7.4 %(V)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00009520
Číslo CAS: 110-54-3
EINECS: 203-777-6
OSN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Assay (GC) Min. 95 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Autoignition Temperature 453 °F
Explosion Limit 7.7 %
Vapor Pressure ~132 mmHg ( 20 °C)
Determination of P/N ≤5 ng/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR