Tlačiť

1-Propanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™

Dodávateľ: Honeywell Chemicals
Nebezpečí

Synonymá: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

33538-1L 33538-6X1L 33538-2.5L 33538-950L
33538-1LEA 114.56 EUR
33538-1L 33538-6X1L 33538-2.5L HONC33538-950L
1-Propanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
1-Propanol

Beilstein Number: 1098242

Vzorec: H₃CCH₂CH₂OH
Mw: 60.1
Teplota varu: 96…98 °C (1.013 hPa)
Teplota tavenia: –127 °C
Hustota: 0.803…0.804 g/cm³ (20 °C)
Teplota vzplanutia: 10.8 %(V)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002941
Číslo CAS: 71-23-8
EINECS: 200-746-9
OSN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Assay (GC) Min. 99.5 %
Boiling range 96 - 99 °C
Density (D 20/20) 0.802 - 0.806
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as C₂H₅COOH) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Acetone (GC) Max. 0.01 %
Ethanol (GC) Max. 0.05 %
Methanol (GC) Max. 0.05 %
2-propanol (GC) Max. 0.05 %
Propionaldehyde (GC) Max. 0.01 %
Autoignition Temperature 700 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR