Tlačiť
VWR® Boiling Stones ('Anti-Bumping'), Granules
Dodávateľ: VWR Chemicals
330093YEA 45.26 EUR
330093Y
VWR® Boiling Stones ('Anti-Bumping'), Granules
Kamínky varné
Granules of fused alumina, to reduce 'bumping' in boiling liquids.
  • Particle size (1-4 mm): min.96%