Tlačiť

Isooktán 99+% ACS

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

Synonymá: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

31787.AP 31787.K2 31787.M1
31787.APEA 57.68 EUR
31787.AP 31787.K2 31787.M1
Isooktán 99+% ACS
Izooktán

Purity: 99+% CAS No.: 540-84-1 Molecular Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 Flash Point: -7°(19°F) Physical Form: Liquid Density: 0.692 Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Solubility: Soluble in benzene, toluene, xylene, chloroform, ether, CS2, CCl4, and DMF Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943


Solvent, thinner, in determination of fuel octane numbers

Vzorec: (CH₃)₂CHCH₂C(CH₃)₃
Mw: 114.23
Teplota varu: 98…99 °C
Teplota tavenia: –107 °C
Hustota: 0.692
Teplota vzplanutia: –7 °C (19 °F)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00008943
Číslo CAS: 540-84-1
EINECS: 208-759-1
OSN: 1262
ADR: 3 II
Merck Index: 14,5193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR