Tlačiť
ExoBrite™ EV Surface Stain Sampler Kit
Dodávateľ: Biotium
30127
30127.EA 590.03 EUR
30127.
ExoBrite™ EV Surface Stain Sampler Kit
Systémy pro spojování a značení proteinů
The ExoBrite™ EV Surface Stain Sampler Kit, Green was developed to offer each of Biotium’s ExoBrite™ EV Surface Stains (CTB, WGA, and Annexin V) for assessing which stain offers the best coverage for the EV samples of interest.