Tlačiť
Reclosable fastener twin pack SJ356D, Dual Lock™
Dodávateľ: 3M
SJ356B
293-0008EA 80.51 EUR
293-0008
Reclosable fastener twin pack SJ356D, Dual Lock™
Pojidla
This reclosable fastener has a clear acrylic adhesive on the back of a clear polypropylene reclosable fastener with 40 stems/cm² and 5,8 mm engaged thickness. This reclosable fastener has a closure life (opened and closed) of 1000 times before losing 50% of its original tensile strength.
  • Snap-in-place installation replaces screws, nails, snaps, and rivets
  • Five times the tensile strength of typical hook-and-loop fasteners
  • Features excellent heat resistance up to temperatures of 93 °C
  • Moisture and UV resistant
  • Suitable for both indoor and outdoor use