Tlačiť

Tetrahydrofuran, Wasser_0,01 99,8 Stabilised, AnalaR NORMAPUR® zA

Dodávateľ: VWR Chemicals
Nebezpečí

Synonymá: Butylene oxide, THF, Diethylene oxide

Stabilizácia: ≤ 300 ppm BHT

28553.293EA 140.6 EUR
28553.293
Tetrahydrofuran, Wasser_0,01 99,8 Stabilised, AnalaR NORMAPUR® zA
Tetrahydrofuran
Vzorec: C₄H₈O
Mw: 72.11
Teplota varu: 66
Teplota tavenia: –108,5 °C
Hustota: 0.888 g/cm³ (20 °C)
Teplota vzplanutia: –21.5 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00005356
Číslo CAS: 109-99-9
EINECS: 203-726-8
OSN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Assay (calculated on dried substance) Min. 99.8 %
Appearance Clear colourless liquid
Acidity Max. 0.0060 %
Peroxides (as H2O2) Max. 0.0200 %
Residue on evaporation Max. 0.0010 %
Stabiliser (Ionol/BHT) Max. 0.030 %
Water Max. 0.0100 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR