Tlačiť

Hydroxid draselný 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Dodávateľ: VWR Chemicals
Nebezpečí

Synonymá: Potash caustic, Caustic potash

26668.263EA 37.09 EUR
26668.263 26668.296 26668.365 26668.460
Hydroxid draselný 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Hydroxid draselný
Vzorec: KOH
Mw: 56.11
Teplota varu: 1320
Teplota tavenia: 360
Hustota: 2.04 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003553
Číslo CAS: 1310-58-3
EINECS: 215-181-3
OSN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Výsledky špecifikačného testu

Assay (calculated as KOH) 85.0 - 100.5 %
Solution in water Passes test
Appearance of solution Passes test Ph.Eur.
Identification B (K) Passes test Ph.Eur.
Solution S1 Passes test Ph.Eur.
Solution S2 Passes test Ph.Eur.
pH (20°C; 0.01 %) Min. 10.5
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 5 ppm
Cl (Chloride) Max. 200 ppm
CO3 (as K2CO3) Max. 1.0 %
PO4 (Phosphate) Max. 5 ppm
SiO4 (as SiO2) Max. 50 ppm
SO4 (Sulphate) Max. 5 ppm
Al (Aluminium) Max. 10 ppm
Ca (Calcium) Max. 10 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 10 ppm
Na (Sodium) Max. 0.5 %
Ni (Nickel) Max. 5 ppm
Pb (Lead) Max. 5 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR