Tlačiť
Scalpel handles, B3/B3L
Dodávateľ: SWANN MORTON
0923
233-5461EA 72.1 EUR
233-5461
Scalpel handles, B3/B3L
Skalpely a skalpelové sady Rukojeti skalpele
Stainless steel, non sterile.
  • Smooth tapered octagonal body offers precise fingertip control
  • Suitable for hand surgery such as a Fasciectomy (removal of scar like tissue ), peripheral nerve repair and surgery of the tendons and joints
  • B3L is longer than the standard B3 providing greater reach and depth of surgical field