Tlačiť
Forceps holder
Dodávateľ: RSG
118.000
232-0189EA 151.01 EUR
232-0189
Forceps holder
Stojany na kleště
Made of stainless steel, ESD coated.
  • For safe storage of all forceps used in the lab
  • Ideal for drying forceps after cleaning
  • Acid resistant, anti-static
  • Can be fastened on tables or under cupboards