Tlačiť
Double spatulas, micro
Dodávateľ: RSG
033.100 033.130 035.150 035.185 035.210 033.150 034.150 034.185 036.150 036.185 036.210 035.130 035.100 033.185 032.150 032.130 032.100 034.100 034.130 034.210 036.130
231-2231EA 34.89 EUR
231-2231 231-2232 231-2233 231-2234 231-2235 231-2236 231-2237 231-0284 231-2239 231-0287 231-2241 231-0467 231-0466 231-0462 231-0461 231-0460 231-0459 231-0463 231-0464 231-0465 231-0468
Double spatulas, micro
Špachtle
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable