Tlačiť
Micro powder spoon
Dodávateľ: RSG
533.005
231-0099EA 25.08 EUR
231-0099
Micro powder spoon
Lžíce
Stainless steel 18/10, anti-magnetic. With hexagonal handle for easier handling.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean