Tlačiť

Dimethylsulfoxid ≥99.9% for HPLC

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

22914.K2 22914.M1
22914.K2EA 246.34 EUR
22914.K2 22914.M1
Dimethylsulfoxid ≥99.9% for HPLC
Dimethylsulfoxid

Purity: 99.9+% CAS No.: 67-68-5 Molecular Formula: (CH3)2SO Formula Weight: 78.13 Flash Point: 87°(189°F) Physical Form: Liquid Density: 1.101 Refractive Index: 1.4790 Sensitivity: Hygroscopic Odour: Mild Melting Point: 18.4° Boiling Point: 189° Beilstein Reference: 506008 Merck Reference: 14,3259 MDL No.: MFCD00002089

Vzorec: (CH₃)₂SO
Mw: 78.14
Teplota varu: 189 °C
Teplota tavenia: 18.4 °C
Hustota: 1.101
Teplota vzplanutia: 87 °C (189 °F)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002089
Číslo CAS: 67-68-5
EINECS: 200-664-3
Merck Index: 14,3259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR