Tlačiť
VWR®, Translucent Euro Containers
Dodávateľ: VWR Collection
216-2447EA 9.17 EUR
216-2447 216-2448 216-2449 216-2450 216-2451 216-2452 216-2453 216-2454
VWR®, Translucent Euro Containers
Krabice Krabice přepravní
These sturdy, transparent plastic containers are made of translucent material. The translucent material allows users to check whether a container is empty or filled, even when stacked or closed.
  • Made of translucent material PPC (polypropylene copolymer)
  • Suitable for manual and automated handling of lightweight goods
  • Smooth surfaces make them easy to clean
  • Ergonomic handles for effective manual handling
  • Suitable for –20 to +65 °C storage
  • 100% recyclable
The translucent Euro containers are available in universal Euros sizes: 300×200, 400×300 and 600×400More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable