Tlačiť
Laboratory trays
Dodávateľ: KARTELL
280 281 282 283 284 285 286 288
216-2010EA 25.2 EUR
216-2010 216-2011 216-2012 216-2013 216-2014 216-2020 216-2025 216-2030
Laboratory trays
Podnosy/Misky/Kelímky Podnosy/misky laboratorní
PVC, white.
  • Thick walled and robust
  • Heat resistant up to 70 °C
  • Smooth surfaces allow easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.