Tlačiť
VWR®, Carboys, Wide Neck, Rectangular
Dodávateľ: VWR Collection
216-1907EA 117.36 EUR
216-1907 216-1898 216-1927
VWR®, Carboys, Wide Neck, Rectangular
Láhve Demižóny
These carboys are useful for storing and dispensing solutions and media.
  • High chemical resistance
  • Ideal for sterile water storage
  • Graduated to identify the volume dispensed
PP carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are for use from –100 to +100 °C.
Note: Always remove the cap from the carboy before autoclaving.
Certifikácie: Resin meets US FDA 21 CFR, USP Class VI.