Tlačiť
VWR®, Ice Pans, with or without Lid
Dodávateľ: VWR Collection
216-1509EA 163.83 EUR
216-1509 216-1525 216-1526 216-1527 216-1528 216-1529 216-1530 216-1531 216-1532 216-1533 216-1537 216-1538 216-1539 216-1493 216-1494 216-1495 216-1496 216-1497 216-1498 216-1499 216-1503 216-1506 216-1507 216-1508
VWR®, Ice Pans, with or without Lid
Nádoby na led
Polyurethane foam.
  • Resistant to temperatures from –20 to +70 °C
  • Extremely robust, leakproof, stackable and lightweight
  • Available in three sizes and four colours
  • Stacking option for space saving on working bench
  • Pouring lip design, for easy draining of liquid
  • Lid to reduce the evaporation level
Ideal for storing samples, these pans maintain hot or cold temperatures of bottles, test tubes, solutions and cultures. Suitable for use with dry ice, water ice and salt solution. For freezer storage only.