Tlačiť
LIGHTBOX Multipurpose Boxes, with Lid
Dodávateľ: MULTIROIR CONTROLEC
LIGHT_BOX_XS LIGHT_BOX_S LIGHT_BOX_M LIGHT_BOX_L LIGHT_BOX_XL
216-1048EA 14.14 EUR
216-1048 216-1049 216-1050 216-1051 216-1052
LIGHTBOX Multipurpose Boxes, with Lid
Krabice Boxy skladovací
Discover this range of polypropylene storage boxes with lids, suitable for storing and organising consumables and laboratory equipment.
  • Available in different sizes
  • Resistant to common chemicals
  • Durable against shocks and frequent handling
  • Stable under temperature variations
  • Secure lids for optimal protection of samples
  • Versatile use for various types of laboratory equipment
  • Easy to clean and decontaminate
Optimise laboratory organisation with this range of polypropylene storage boxes, specifically designed to meet the demands of scientificMore Product Information