Tlačiť
Test tube racks, radioimmunassay, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F188720000 F188710000 F188700000
212-6501EA 236.73 EUR
212-6501 212-6502 212-6503
Test tube racks, radioimmunassay, SP Bel-Art
Stojany Tube Racks
PP, white.
  • Temperature resistant to 121 °C, steam sterilisable
  • Good chemical resistance
These racks provide a practical method of pipetting and storing a large number of samples. By using alternate rows and rotating the tubes, it is easy to keep track of the last tube filled. Also useful in clinical laboratories for storage of samples.