Tlačiť
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Dodávateľ: EPPENDORF
0030125.150 0030125.177 0030125.185 0030125.193 0030125.207 0030125.215
700-5239EA 51.67 EUR
700-5239 211-2114 211-2116 211-2117 211-2118 211-2166
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Zkumavky Mikrozkumavky centrifugační
Eppendorf Tubes® 3810X microtubes (1,5 ml) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Perfect for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into workflows for efficient and reliable performance.
  • Easy-to-use ergonomic design featuring a user-friendly tube lid
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures
  • g-Safe® technology ensures stability up to 30000×g, preventing sample loss during
More Product Information