Tlačiť

Kyselina sírová 95,0 - 97,0% 95,0-97,0, AnalaR NORMAPUR® zA

Dodávateľ: VWR Chemicals
Nebezpečí

20700.265EA 34.97 EUR
20700.265 20700.298 20700.243 20700.323 20700.420 20700.290 20700.320 20700.460 20700.550
Kyselina sírová 95,0 - 97,0% 95,0-97,0, AnalaR NORMAPUR® zA
Kyselina sírová
Vzorec: H₂SO₄
Mw: 98.08
Teplota varu: 330 °C (1013 hPa)
Teplota tavenia: 10.38 °C
Hustota: 1.84 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00064589
Číslo CAS: 7664-93-9
EINECS: 231-639-5
OSN: 1830
ADR: 8 II
REACH: 01-2119458838-20

Order Now

Výsledky špecifikačného testu

Assay 95.0 - 97.0 %
Colouration Max. 10 APHA
Ignition residue Max. 5 ppm
Substances reducing KMnO4 (as SO2) Max. 2 ppm
Cl (Chloride) Max. 0.2 ppm
NH4 (Ammonium) Max. 2 ppm
NO3 + NO2 (as NO3) Max. 0.2 ppm
PO4 (Phosphate) Max. 0.5 ppm
Ag (Silver) Max. 0.02 ppm
Al (Aluminium) Max. 0.02 ppm
As (Arsenic) Max. 0.005 ppm
Ba (Barium) Max. 0.02 ppm
Be (Beryllium) Max. 0.01 ppm
Bi (Bismuth) Max. 0.01 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.02 ppm
K (Potassium) Max. 0.05 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.03 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.02 ppm
Ti (Titanium) Max. 0.02 ppm
Tl (Thallium) Max. 0.02 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.1 ppm
Zr (Zirconium) Max. 0.02 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR